Skip to Content
Merck
All Photos(1)

Key Documents

779377

Sigma-Aldrich

1-Azido-4-bromobenzene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

4-Bromophenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrN3
CAS Number:
Molecular Weight:
198.02
Beilstein:
1939308
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Brc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C6H4BrN3/c7-5-1-3-6(4-2-5)9-10-8/h1-4H

InChI key

WHSJSMSBFMDFHK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service