Skip to Content
Merck
All Photos(1)

Documents

757772

Sigma-Aldrich

Poly(ethylene glycol) 2-aminoethyl ether biotin

average Mn 5,300, cross-linking reagent carboxyl reactive, amine, biotin

Synonym(s):

Polyethylene glycol, aminoPEG biotin, biotin polyoxyethylene amine, biotin-PEG, biotin-PEG-amine, biotinylated PEG, polyoxyethylene ethylamine ether biotin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) 2-aminoethyl ether biotin, average Mn 5,300

form

solid

Quality Level

mol wt

PEG average Mn 5,000 (n~110)
average Mn 5,300

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

mp

52-57 °C

Ω-end

amine

α-end

biotin

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Biotin-PEG-amine may be used for the biotinylation reaction of polymeric worm micelles for easy targeting and drug transfer to cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Targeted worm micelles.
Dalhaimer P, et al.
Biomacromolecules, 5(5), 1714-1719 (2004)

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service