Skip to Content
Merck
All Photos(1)

Key Documents

745472

Sigma-Aldrich

cis-9-Tetradecenoyl-L-carnitine hydrochloride

97% (CP)

Synonym(s):

cis-9-Tetradecenoyl-L-carnitine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H39NO4 · xHCl
Molecular Weight:
369.54 (free base basis)
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

97% (CP)

form

solid

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](CC([O-])=O)OC(CCCCCCC/C=C\CCCC)=O.Cl

InChI

1S/C21H39NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4;/h8-9,19H,5-7,10-18H2,1-4H3;1H/b9-8-;/t19-;/m0./s1

InChI key

CNZGWHVCFLOOMJ-BVNCMAKQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service