Skip to Content
Merck
All Photos(1)

Key Documents

724122

Sigma-Aldrich

2-Bromo-4-tert-butylthiazole

97%

Synonym(s):

2-Bromo-4-(1,1-dimethylethyl)thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10BrNS
CAS Number:
Molecular Weight:
220.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.539

density

1.383 g/mL at 25 °C

SMILES string

CC(C)(C)c1csc(Br)n1

InChI

1S/C7H10BrNS/c1-7(2,3)5-4-10-6(8)9-5/h4H,1-3H3

InChI key

ILKZEOZQPGBUCV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.0 °F

Flash Point(C)

98.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service