714690
Ammonium formate solution
10 mM in H2O, suitable for HPLC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HCOONH4
CAS Number:
Molecular Weight:
63.06
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
Recommended Products
grade
HPLC grade
form
liquid
concentration
10 mM in H2O
technique(s)
HPLC: suitable
mp
-119-121 °C (lit.)
density
0.998 g/mL at 25 °C
application(s)
food and beverages
SMILES string
[NH4+].[O-]C=O
InChI
1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3
InChI key
VZTDIZULWFCMLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ammonium formate solution may be used to prepare the background electrolyte (BGE) for the enantiomeric separation of salbutaminol by nonaqueous capillary electrophoresis (NACE) with electrospray ionization (ESI) mass spectrometry (MS).
Packaging
M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
219.2 °F
Flash Point(C)
104 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
On-line coupling of cyclodextrin mediated nonaqueous capillary electrophoresis to mass spectrometry for the determination of salbutamol enantiomers in urine.
Servais AC, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(3), 752-757 (2006)
Anne M Filppula et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(7), 1202-1209 (2014-04-10)
Previous studies have shown that several protein kinase inhibitors are time-dependent inhibitors of cytochrome P450 (CYP) 3A. We screened 14 kinase inhibitors for time-dependent inhibition of CYP2C8 and CYP3A. Amodiaquine N-deethylation and midazolam 1'-hydroxylation were used as marker reactions for
Therese Fagerqvist et al.
Journal of neurochemistry, 126(1), 131-144 (2013-02-01)
Inclusions of intraneuronal alpha-synuclein (α-synuclein) can be detected in brains of patients with Parkinson's disease and dementia with Lewy bodies. The aggregation of α-synuclein is a central feature of the disease pathogenesis. Among the different α-synuclein species, large oligomers/protofibrils have
Jo Philips et al.
Journal of contaminant hydrology, 147, 25-33 (2013-03-19)
Acidification due to microbial dechlorination of trichloroethene (TCE) can limit the bio-enhanced dissolution of TCE dense non-aqueous phase liquid (DNAPL). This study related the dissolution enhancement of a TCE DNAPL to the pH buffer capacity of the medium and the
Lucie Nováková et al.
Analytica chimica acta, 853, 637-646 (2014-12-04)
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service