689122
O,O′-1,3-Propanediylbishydroxylamine dihydrochloride
98%
Synonym(s):
O,O′-Diaminopropane-1,3-diol dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H10N2O2 · 2HCl
CAS Number:
Molecular Weight:
179.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
crystals
impurities
≤1.0% water
SMILES string
NOCCCON.[H]Cl.[H]Cl
InChI
1S/2ClH/h2*1H
InChI key
IXCSERBJSXMMFS-UHFFFAOYSA-N
Application
Reactant for preparation of:
- Dinitrobenzamide-TEMPO derivatives
- Oxa-azamacrocycles
- Adamantylalkoxyurea derivatives
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service