Skip to Content
Merck
All Photos(1)

Key Documents

664960

Sigma-Aldrich

3-Chloro-1-phenyl-1-propyne

97%

Synonym(s):

1-Chloro-3-phenyl-2-propyne, 3-Phenylpropargyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl
CAS Number:
Molecular Weight:
150.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.585

density

1.095 g/mL at 25 °C

functional group

chloro
phenyl

SMILES string

ClCC#Cc1ccccc1

InChI

1S/C9H7Cl/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,8H2

InChI key

RMPSZEZJKSUNKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.9 °F

Flash Point(C)

104.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service