655031
2-Methoxyphenethyl bromide
97%
Synonym(s):
2-Methoxy-1-(2-bromoethyl)benzene
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CH3OC6H4CH2CH2Br
Molecular Weight:
215.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.558 (lit.)
bp
105.5-110.5 °C/5 mmHg (lit.)
density
1.356 g/mL at 25 °C (lit.)
functional group
bromo
SMILES string
COc1ccccc1CCBr
InChI
1S/C9H11BrO/c1-11-9-5-3-2-4-8(9)6-7-10/h2-5H,6-7H2,1H3
InChI key
UIIJKBSUGZNVQN-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service