Skip to Content
Merck
All Photos(1)

Key Documents

649171

Sigma-Aldrich

Poly(2-propylacrylic acid)

average Mn 20,000-80,000

Synonym(s):

2-Propylacrylic acid homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

mol wt

average Mn 20,000-80,000

storage temp.

2-8°C

SMILES string

OC(=O)C(=C)CCC

InChI key

HEBDGRTWECSNNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

pH-sensitive polymer for drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service