Skip to Content
Merck
All Photos(1)

Key Documents

615838

Sigma-Aldrich

2-Methylbutane-d12

98 atom % D

Synonym(s):

Isopentane-d12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2CDCD2CD3
CAS Number:
Molecular Weight:
84.22
MDL number:
UNSPSC Code:
12352202

isotopic purity

98 atom % D

Quality Level

mass shift

M+12

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyril Andrieu et al.
Development (Cambridge, England), 147(7) (2020-04-14)
Matrix metalloproteinases have a broad spectrum of substrates ranging from extracellular matrix components and adhesion molecules to chemokines and growth factors. Despite being mostly secreted, MMPs have been detected in the cytosol, the mitochondria or the nucleus. Although most of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service