Skip to Content
Merck
All Photos(1)

Key Documents

492256

Sigma-Aldrich

Dimethyl phthalate-3,4,5,6-d4

98 atom % D

Synonym(s):

1,2-Dimethyl 1,2-benzene-3,4,5,6-d4-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4(CO2CH3)2
CAS Number:
Molecular Weight:
198.21
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.214 g/mL at 25 °C

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OC)c(c1[2H])C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D

InChI key

NIQCNGHVCWTJSM-LNFUJOGGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This product may be used as an analytical standard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service