Skip to Content
Merck
All Photos(1)

Key Documents

491675

Sigma-Aldrich

Acetonitrile-13C2,15N

98 atom % 15N, 99 atom % 13C

Synonym(s):

Methyl cyanide-13C2,15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313C15N
CAS Number:
Molecular Weight:
44.03
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

99% (CP)

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−48 °C (lit.)

density

0.843 g/mL at 25 °C

mass shift

M+3

SMILES string

[13CH3][13C]#[15N]

InChI

1S/C2H3N/c1-2-3/h1H3/i1+1,2+1,3+1

InChI key

WEVYAHXRMPXWCK-VMIGTVKRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service