482374
Poly(dimethylsiloxane-co-methylhydrosiloxane), trimethylsilyl terminated
average Mn ~13,000, methylhydrosiloxane 3-4 mol %
About This Item
Recommended Products
mol wt
average Mn ~13,000
composition
methylhydrosiloxane, 3-4 mol %
refractive index
n20/D 1.405
viscosity
~260 cSt(lit.)
bp
>93 °C (lit.)
density
0.97 g/mL at 25 °C
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
150.1 °F - closed cup
Flash Point(C)
65.6 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.
Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.
Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.
Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.
Related Content
We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.
We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.
We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.
We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service