Skip to Content
Merck
All Photos(3)

Key Documents

460028

Sigma-Aldrich

(R)-(−)-2,2-Dimethyl-5-oxo-1,3-dioxolane-4-acetic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O5
CAS Number:
Molecular Weight:
174.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]20/D −3.5°, c = 1 in chloroform

mp

108-112 °C (lit.)

SMILES string

CC1(C)O[C@H](CC(O)=O)C(=O)O1

InChI

1S/C7H10O5/c1-7(2)11-4(3-5(8)9)6(10)12-7/h4H,3H2,1-2H3,(H,8,9)/t4-/m1/s1

InChI key

IDQOCLIWDMZKBZ-SCSAIBSYSA-N

General description

(R)-(−)-2,2-Dimethyl-5-oxo-1,3-dioxolane-4-acetic acid can be used as an intermediate/starting material in the synthesis of (R)-isoserine, a glucagon receptor antagonist, blepharismone, and pheromone of Blepharisma japonicum.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel glucagon receptor antagonists with improved selectivity over the glucose-dependent insulinotropic polypeptide receptor
Kodra JT, et al.
Journal of medicinal chemistry, 51(17), 5387-5396 (2008)
Practical synthesis of blepharismone, a mating inducing pheromone of Blepharisma japonicum
Takihiro H, et al.
Tetrahedron Asymmetry, 17(16), 2339-2343 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service