Skip to Content
Merck
All Photos(1)

Documents

451053

Sigma-Aldrich

Bis(dichlorophosphino)methane

95%

Synonym(s):

Methylenebis(dichlorophosphine), Methylenebis(phosphonous dichloride)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2PCH2PCl2
CAS Number:
Molecular Weight:
217.79
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.5940 (lit.)

density

1.601 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

ClP(Cl)CP(Cl)Cl

InChI

1S/CH2Cl4P2/c2-6(3)1-7(4)5/h1H2

InChI key

OEKLCAXLANZKPX-UHFFFAOYSA-N

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

205.0 °F - closed cup

Flash Point(C)

96.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service