Skip to Content
Merck
All Photos(2)

Key Documents

440965

Sigma-Aldrich

Poly(1,4-phenylene ether-ether-sulfone)

pellets

Synonym(s):

4,4′-Dichlorodiphenyl sulfone-p -hydroquinone copolymer, SRU, Dichlorodiphenyl sulfone-hydroquinone copolymer, SRU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4-4-SO2C6H4-4-OC6H4-4-O)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

pellets

transition temp

Tg 192 °C

SMILES string

Oc1ccc(O)cc1.Oc2ccc(cc2)S(=O)(=O)c3ccc(O)cc3

InChI

1S/C12H10O4S.C6H6O2/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12;7-5-1-2-6(8)4-3-5/h1-8,13-14H;1-4,7-8H

InChI key

AEAZYDFBOSHKHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service