Skip to Content
Merck
All Photos(1)

Key Documents

438197

Sigma-Aldrich

Poly(ethylene glycol-ran-propylene glycol)

Mn ~2,500

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

Mn ~2,500

refractive index

n20/D 1.465

viscosity

650 cSt(25 °C)(lit.)

bp

>200 °C (lit.)

transition temp

pour point 4 °C

solubility

H2O: soluble at <70 °C

density

1.095 g/mL at 25 °C

SMILES string

OCCO.CC(O)CO

Looking for similar products? Visit Product Comparison Guide

Application

Hydraulic fluids, metal working fluids and lubricants, heat transfer fluids, solder assist fluids, quenchants, lubricants, solvents, plasticizers and foam control agents.

Features and Benefits

Good lubricity under load, noncorrosive to metals. Very low ash content, low pour point, excellent thermal stability.

Physical form

Dihydroxy functional.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service