Skip to Content
Merck
All Photos(2)

Key Documents

432415

Sigma-Aldrich

Polystyrene-block-polyisoprene-block-polystyrene

styrene 22 wt. %

Synonym(s):

Polystyrene-block-polyisoprene, multi-arm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]X[CH2CH=C(CH3)CH2]Y[CH2CH(C6H5)]z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

melt index

3 g/10 min (200°C/5.0kg)

contains

phenolic as antioxidant

composition

styrene, 22 wt. %

hardness

52 (Shore A)

viscosity

12 poise, 25 wt. % in toluene(25 °C, Brookfield)(lit.)

solubility

solvents with solubility parameters between 7.7 and 9.4: soluble

density

0.93 g/mL at 25 °C

elongation

ASTM D 412 - 1,200% (ultimate)

SMILES string

CC(=C)C=C.C=Cc1ccccc1

InChI

1S/C8H8.C5H8/c1-2-8-6-4-3-5-7-8;1-4-5(2)3/h2-7H,1H2;4H,1-2H2,3H3

InChI key

ROGIWVXWXZRRMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Component of hot melt adhesives, sealants, asphalt and oil gels. Modifier for polymers, thermosets and general rubber compounding. Direct molded or extruded auto parts, sporting goods, footware and film.

Features and Benefits

High strength and low viscosity for easy melt and solution processing. High temperature degradation results in cross-linking. Flexible to -65°C. Excellent electrical insulator, resists acids and alkali.

Physical form

Thermoplastic elastomer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sneha B Chakrapani et al.
The Analyst, 144(5), 1679-1686 (2019-01-19)
The accurate characterization of relative composition in multicomponent polymer systems such as statistical copolymers, block copolymers, and polymer blends is critical to understanding and predicting their behavior. Typically, polymer compositional analysis is performed using 1H Nuclear Magnetic Resonance (NMR) Spectroscopy
Nanjia Zhou et al.
Science advances, 5(5), eaav8141-eaav8141 (2019-06-07)
One-dimensional (1D) nanomaterials with highly anisotropic optoelectronic properties are key components in energy harvesting, flexible electronics, and biomedical imaging devices. 3D patterning methods that precisely assemble nanowires with locally controlled composition and orientation would enable new optoelectronic device designs. As

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service