Skip to Content
Merck
All Photos(1)

Documents

404438

Sigma-Aldrich

(S,S)-(+)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine

98%

Synonym(s):

(S,S)-Jacobsen’s ligand

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3C]2C6H2-2-(OH)CH=N]2C6H10
CAS Number:
Molecular Weight:
546.83
Beilstein:
5464823
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]20/D +315 to 15°, c = 1 in methylene chloride

mp

203-206 °C (lit.)

SMILES string

CC(C)(C)c1cc(\C=N\[C@H]2CCCC[C@@H]2\N=C\c3cc(cc(c3O)C(C)(C)C)C(C)(C)C)c(O)c(c1)C(C)(C)C

InChI

1S/C36H54N2O2/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12/h17-22,29-30,39-40H,13-16H2,1-12H3/b37-21+,38-22+/t29-,30-/m0/s1

InChI key

FYNXDGNCEBQLGC-GTHUDTLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S,S)-(+)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine is a chiral salen-type ligand used for preparing Jacobsen′s catalyst suitable for enantioselective epoxidation of olefins lacking functional groups.

Legal Information

Licensed through Research Corporation Technologies

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang, S. et al.
Journal of the American Chemical Society, 58, 6939-6939 (1993)
E.N. Jacobsen et al.
Journal of the American Chemical Society, 113, 7063-7063 (1991)
L. Deng et al.
The Journal of Organic Chemistry, 57, 4320-4320 (1992)

Articles

The first reports of salen used as ligands with manganese for asymmetric epoxidation reactions.

The first reports of salen used as ligands with manganese for asymmetric epoxidation reactions.

The first reports of salen used as ligands with manganese for asymmetric epoxidation reactions.

The first reports of salen used as ligands with manganese for asymmetric epoxidation reactions.

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service