Skip to Content
Merck
All Photos(1)

Key Documents

367702

Sigma-Aldrich

Thulium(III) acetate hydrate

99.9% trace metals basis

Synonym(s):

Thulium(III) acetic acid salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3Tm · xH2O
CAS Number:
Molecular Weight:
346.07 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.9% trace metals basis

form

powder

reaction suitability

core: thulium
reagent type: catalyst

SMILES string

O.CC(=O)O[Tm](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.H2O.Tm/c3*1-2(3)4;;/h3*1H3,(H,3,4);1H2;/q;;;;+3/p-3

InChI key

GKCQSJXCECKBHO-UHFFFAOYSA-K

General description

Thulium(III) acetate hydrate is a chemical compound containing the rare earth element thulium. It can be used as a precursor in the synthesis of core-shell nanocrystals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of core-shell lanthanide-doped upconversion nanocrystals for cellular applications
Ai X, et al.
Journal of Visualized Experiments, 129, e56416-e56416 (2017)
Yuejiao Xian et al.
PloS one, 15(4), e0221180-e0221180 (2020-04-23)
At the molecular level, the circadian clock is regulated by a time delayed transcriptional-translational feedback loop in which the core proteins interact with each other rhythmically to drive daily biological rhythms. The C-terminal domain of a key clock protein PER2
Qianqian Su et al.
Frontiers in chemistry, 8, 836-836 (2020-10-24)
Lanthanide-based upconversion nanoparticles can convert low-energy excitation to high-energy emission. The self-assembled upconversion nanoparticles with unique structures have considerable promise in sensors and optical devices due to intriguing properties. However, the assembly of isotropic nanocrystals into anisotropic structures is a
Qiyue Shao et al.
Nanoscale, 9(33), 12132-12141 (2017-08-15)
Upconversion nanoparticles (UCNPs) are an excellent choice to construct security features against counterfeiting, owing to their unique NIR-to-VIS upconversion luminescence (UCL) characteristics. However, the application of upconversion materials is limited, due to their single and invariant emission colors. Herein, the
Shihua Li et al.
ACS nano, 13(2), 2103-2113 (2019-01-16)
The exploitation of gas therapy platforms holds great promise as a "green" approach for selective cancer therapy, however, it is often associated with some challenges, such as uncontrolled or insufficient gas generation and unclear therapeutic mechanisms. In this work, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service