Skip to Content
Merck
All Photos(2)

Documents

340642

Sigma-Aldrich

Cellulose acetate propionate

average Mn ~15,000 by GPC, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H114O49
CAS Number:
Molecular Weight:
1811.69
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn ~15,000 by GPC

extent of labeling

0.0-1.0 wt. % Acetyl
~5 wt. % Hydroxyl
40-45 wt. % Propionyl

mp

188-210 °C (lit.)

density

1.23 g/mL at 25 °C (lit.)

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O.CC(=O)OC[C@H]3O[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]3O[C@@H]4O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]4OC(C)=O.CCC(=O)OC[C@H]5O[C@@H](OC(=O)CC)[C@H](OC(=O)CC)[C@@H](OC(=O)CC)[C@@H]5O[C@@H]6O[C@H](COC(=O)CC)[C@@H](OC(=O)CC)[C@H](OC(=O)CC)[C@H]6OC(=O)CC

InChI

1S/C36H54O19.C28H38O19.C12H22O11/c1-9-21(37)45-17-19-29(49-23(39)11-3)31(50-24(40)12-4)34(53-27(43)15-7)36(48-19)55-30-20(18-46-22(38)10-2)47-35(54-28(44)16-8)33(52-26(42)14-6)32(30)51-25(41)13-5;1-11(29)37-9-19-21(39-13(3)31)23(40-14(4)32)26(43-17(7)35)28(46-19)47-22-20(10-38-12(2)30)45-27(44-18(8)36)25(42-16(6)34)24(22)41-15(5)33;13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h19-20,29-36H,9-18H2,1-8H3;19-28H,9-10H2,1-8H3;3-20H,1-2H2/t19-,20-,29-,30-,31+,32+,33-,34-,35+,36+;19-,20-,21-,22-,23+,24+,25-,26-,27-,28+;3-,4-,5-,6+,7-,8-,9-,10-,11-,12+/m111/s1

InChI key

HKQOBOMRSSHSTC-DIBAFDJWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S B Bhardwaj et al.
Journal of microencapsulation, 12(1), 71-81 (1995-01-01)
Microspheres containing two different drug loadings of a calcium channel blocker, verapamil hydrochloride, were prepared with three different cellulose esters namely cellulose acetate (CA), cellulose acetate propionate (CAP) and cellulose acetate butyrate (CAB) of approximately similar molecular weights using the
M Efentakis et al.
International journal of pharmaceutics, 311(1-2), 147-156 (2006-01-27)
In this investigation a novel oral pulsatile drug delivery system based on a core-in-cup dry coated tablet, where the core tablet surrounded on the bottom and circumference wall with inactive material, is proposed. The system consists of three different parts
A J Shukla et al.
Pharmaceutical research, 8(11), 1396-1400 (1991-11-01)
Microspheres with 40, 50, and 60% drug loading of anhydrous theophylline core material were prepared by the emulsion-solvent evaporation method. Three different molecular weights of cellulose acetate propionate were used as encapsulating polymers. The geometric mean diameter of the microspheres
Kelin Huang et al.
Journal of agricultural and food chemistry, 59(10), 5376-5381 (2011-04-02)
Cellulose acetate butyrate (CAB) and cellulose acetate propionate (CAP) were prepared homogeneously in a 1-allyl-3-methylimidazolium chloride (AmimCl) ionic liquid system from sugarcane bagasse (SB). The reaction temperature, reaction time, and molar ratio of butyric (propionic) anhydride/anhydroglucose units in the cellulose
Manuela C C M Sobral et al.
Journal of materials science. Materials in medicine, 19(2), 677-682 (2007-07-11)
Ketotifen was immobilised in cellulose acetate propionate (CAP) membranes and in cellulose acetate butyrate (CAB) membranes. The characteristics of each system were evaluated under a range of experimental conditions. The topography and uniformity of the membranes was assessed using scanning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service