Skip to Content
Merck
All Photos(1)

Key Documents

287628

Sigma-Aldrich

Ethyl difluoroacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2CHCO2C2H5
CAS Number:
Molecular Weight:
124.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.347 (lit.)

bp

99.2 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)C(F)F

InChI

1S/C4H6F2O2/c1-2-8-4(7)3(5)6/h3H,2H2,1H3

InChI key

GZKHDVAKKLTJPO-UHFFFAOYSA-N

General description

Ethyl difluoroacetate was degraded to difluoroacetic acid by the strains of Actinobacteria[1].

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F - closed cup

Flash Point(C)

23.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening of fluorinated materials degrading microbes.
Iwai N, et al.
Journal of Fluorine Chemistry, 130(4), 434-437 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service