Skip to Content
Merck
All Photos(1)

Documents

277134

Sigma-Aldrich

Sodium biphenyl complex

in 2-ethoxyethyl ether

Synonym(s):

Biphenyl sodium complex solution, Sodium biphenyl complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5 · Na
CAS Number:
Molecular Weight:
177.20
Beilstein:
3989646
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.6 (vs air)

Quality Level

form

liquid

reaction suitability

reagent type: reductant

packaging

pk of 20 × 15 mL

concentration

in 2-ethoxyethyl ether

density

1.1 g/mL at 25 °C

SMILES string

[Na]c1ccc(cc1)-c2ccccc2

InChI

1S/C12H9.Na/c1-3-7-11(8-4-1)12-9-5-2-6-10-12;/h1,3-10H;

InChI key

UUNVZHGYJSUMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium biphenyl reagent is used to cleave the covalent fluorine bonds in organic compounds and biological samples. The liberated fluoride ions are determined spectrophotometrically or with the fluoride ion electrode.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of organically bound fluorine using sodium biphenyl reagent and a fluoride?specific ion electrode.
Jones BC, et al.
Journal of Pharmaceutical Sciences, 60(7), 1036-1039 (1971)
Sodium biphenyl method for determination of covalently bound fluorine in organic compounds and biological materials.
Venkateswarlu P.
Analytical Chemistry, 54(7), 1132-1137 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service