Skip to Content
Merck
All Photos(1)

Key Documents

274402

Sigma-Aldrich

Tetramethyltetraselenafulvalene

97%

Synonym(s):

TMTSF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12Se4
CAS Number:
Molecular Weight:
448.04
Beilstein:
1619644
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

273-275 °C (dec.) (lit.)

SMILES string

CC1=C(C)[Se]\C([Se]1)=C2\[Se]C(C)=C(C)[Se]2

InChI

1S/C10H12Se4/c1-5-6(2)12-9(11-5)10-13-7(3)8(4)14-10/h1-4H3

InChI key

YMWLPMGFZYFLRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethyltetraselenafulvalene salts and charge-transfer complexes exhibit superconducting properties.

Application

Tetramethyltetraselenafulvalene was used in the preparation of charge-transfer salt nanowires using the galvanostatic deposition technique.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youn Jung Jung et al.
Journal of nanoscience and nanotechnology, 12(7), 5397-5401 (2012-09-13)
TMTSF-based (TMTSF = tetramethyltetraselenafulvalene = C10H12Se4) charge-transfer salt nanowires were fabricated using the galvanostatic deposition technique that was assisted by an anodic aluminum oxide (AAO) template. By applying a low current density of 1-2 microA/cm2 for more than one month
Front. Solid State Sci., 1, 81-81 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service