Skip to Content
Merck
All Photos(2)

Documents

245682

Sigma-Aldrich

3-Chlorophenyl isocyanate

99%

Synonym(s):

3-Isocyanato chlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NCO
CAS Number:
Molecular Weight:
153.57
Beilstein:
742445
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.559 (lit.)

bp

113-114 °C/43 mmHg (lit.)

mp

−4 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

Clc1cccc(c1)N=C=O

InChI

1S/C7H4ClNO/c8-6-2-1-3-7(4-6)9-5-10/h1-4H

InChI key

HHIRBXHEYVDUAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chlorophenyl isocyanate (3-isocyanato chlorobenzene) was used in the preparation of cis-4-[N-(3-chlorophenyl)carbamoloxyl]-2-butenyl-trimethylammoniumiodide.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereochemical analogs of a muscarinic, ganglionic stimulant. cis- and trans-4-(N-(3-chlorophenyl)carbamoyloxy)-2-butenyltrimethylammonium iodides.
W L Nelson et al.
Journal of medicinal chemistry, 16(5), 506-510 (1973-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service