Skip to Content
Merck
All Photos(1)

Documents

225177

Sigma-Aldrich

Trimesitylborane

97%

Synonym(s):

NSC 149907, Tris(2,4,6-trimethylphenyl) borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[2,4,6-(CH3)3C6H2]3B
CAS Number:
Molecular Weight:
368.36
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

193-195 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(B(c2c(C)cc(C)cc2C)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C27H33B/c1-16-10-19(4)25(20(5)11-16)28(26-21(6)12-17(2)13-22(26)7)27-23(8)14-18(3)15-24(27)9/h10-15H,1-9H3

InChI key

FXFWLXFJBVILBE-UHFFFAOYSA-N

Application

Reactant involved in:
  • Synthesis of sandwich complex of trimesitylborane and fluoroborate and cyanoborate derivatives
  • Investigations of reactivity of aryldimesitylboranes under Suzuki-Miyaura coupling conditions
  • Laser desorption ionization mass spectrometry investigations
  • Studies of ion conductive characteristics of boron stabilized carbanions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service