Skip to Content
Merck
All Photos(2)

Key Documents

209031

Sigma-Aldrich

Di-μ-chloro-tetracarbonyldirhodium(I)

97%

Synonym(s):

Rhodium carbonyl chloride(I), Rhodium(I) carbonyl chloride, Rhodium(I) dicarbonyl chloride dimer, Tetracarbonyldi-μ−chlorodirhodium(I)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Rh(CO)2Cl]2
CAS Number:
Molecular Weight:
388.76
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

mp

120-125 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cl+]1[Rh-][Cl+][Rh-]1

InChI

1S/4CO.2Cl.2Rh/c4*1-2;;;;/q;;;;2*+1;2*-1

InChI key

ODYPMWFHWPMOKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di-μ-chloro-tetracarbonyldirhodium(I) can be used as a catalyst or catalyst precursor for various organic reactions such as Pauson-Khand reaction, allylic alkylations, rearrangement of strained rings, carbonylative ring expansion, carbonylative coupling of organometallics, hydroformylation, silylformylation, and cycloaddition reactions of alkenes, alkynes, and allenes.

Application

  • Rhodium-Catalyzed Trans-Bis-Silylation Reactions: This study explores the use of Di-μ-chloro-tetracarbonyldirhodium(I) as a catalyst in the synthesis of specialized pyridine derivatives, demonstrating potential for creating complex molecular architectures useful in drug discovery and material science (Naka & Kobayashi, 2023).
  • Redox behaviour of imino-β-diketonato ligands: The research investigates the coordination chemistry of Di-μ-chloro-tetracarbonyldirhodium(I) with innovative ligands, potentially opening new pathways for catalytic systems used in pharmaceutical syntheses (Ferreira, Conradie & Conradie, 2022).
  • Synthesis of Thiophene-Fused Siloles through Rhodium-Catalyzed Trans-Bis-Silylation: The application of Di-μ-chloro-tetracarbonyldirhodium(I) in the synthesis of thiophene-fused siloles indicates its role in advancing materials chemistry for electronic and photonic applications (Naka et al., 2024).
  • Kinetic Study of the Oxidative Addition Reaction between Methyl Iodide and Complexes: Utilizing UV–Vis, IR spectroscopy, this study enhances understanding of the mechanisms involved when using Di-μ-chloro-tetracarbonyldirhodium(I) in catalytic cycles, relevant to both academia and industry settings (Ferreira et al., 2022).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 4187-4187 (1993)
Wender, P.A. Sperandio, D.
The Journal of Organic Chemistry, 63, 4164-4164 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service