Skip to Content
Merck
All Photos(1)

Documents

181919

Sigma-Aldrich

Poly(ethylene adipate)

average Mw ~10,000 by GPC

Synonym(s):

Ethylene glycol adipate, Poly(ethylene adipate), dihydroxy terminated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[OCH2CH2O2C(CH2)4CO]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

chunks

Quality Level

mol wt

average Mw ~10,000 by GPC

density

1.183 g/mL at 25 °C

InChI

1S/C6H10O4.C2H5O2/c7-5(8)3-1-2-4-6(9)10;3-1-2-4/h1-4H2,(H,7,8)(H,9,10);3H,1-2H2

InChI key

CODQTHDBFINLJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service