Skip to Content
Merck
All Photos(1)

Key Documents

175919

Sigma-Aldrich

m-Xylene-d10

98 atom % D

Synonym(s):

1,3-Dimethylbenzene-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4(CD3)2
CAS Number:
Molecular Weight:
116.23
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

technique(s)

GC/MS: suitable
NMR: suitable

impurities

≤0.001% water
water

refractive index

n20/D 1.497 (lit.)

bp

138-139 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c(c([2H])c(c1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3/i1D3,2D3,3D,4D,5D,6D

InChI key

IVSZLXZYQVIEFR-ZGYYUIRESA-N

General description

m-Xylene-d10 is a deuterated derivative of m-xylene. It has an isotopic purity of 99atom%D. The rotational spectra (range of 6 and 26.5GHz) and rotational constants of m-xylene and m-xylene-d10 have been evaluated by pulsed beam Fourier transform microwave spectroscopy.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Microwave Spectra of m-Xylene and m-Xylene-d10. Determination of the Low Methyl Internal Rotation Barrier.
Thomsen C and Dreizler H.
Zeitschrift fur Naturforschung, A, 56(9-10), 635-640 (2001)
F Aladar Bencsath et al.
Analytical and bioanalytical chemistry, 407(14), 4079-4090 (2015-03-23)
A headspace solid-phase microextraction gas chromatography-mass spectrometry (SPME GC-MS) method is described, to screen seafood for volatile organic compounds (VOCs) associated with petrochemical taint. VOCs are extracted from the headspace of heated sample homogenates by adsorption onto a SPME fiber

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service