Skip to Content
Merck
All Photos(1)

Key Documents

16599

Sigma-Aldrich

4-(Trifluoromethyl)-L-phenylglycine

≥98.0%

Synonym(s):

(S)-2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8F3NO2
CAS Number:
Molecular Weight:
219.16
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

N[C@H](C(O)=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C9H8F3NO2/c10-9(11,12)6-3-1-5(2-4-6)7(13)8(14)15/h1-4,7H,13H2,(H,14,15)/t7-/m0/s1

InChI key

FANMQHUKZBBELZ-ZETCQYMHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingliang Zhang et al.
Cell reports, 36(5), 109495-109495 (2021-08-05)
Scn2a encodes the voltage-gated sodium channel NaV1.2, a main mediator of neuronal action potential firing. The current paradigm suggests that NaV1.2 gain-of-function variants enhance neuronal excitability, resulting in epilepsy, whereas NaV1.2 deficiency impairs neuronal excitability, contributing to autism. However, this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service