Skip to Content
Merck
All Photos(1)

Key Documents

114154

Sigma-Aldrich

trans-3,6-Endomethylene-1,2,3,6-tetrahydrophthaloyl chloride

97%

Synonym(s):

trans-5-Norbornene-2,3-dicarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl2O2
CAS Number:
Molecular Weight:
219.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.517 (lit.)

bp

114-118 °C/11 mmHg (lit.)

density

1.349 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)[C@@H]1[C@@H]2C[C@@H](C=C2)[C@H]1C(Cl)=O

InChI

1S/C9H8Cl2O2/c10-8(12)6-4-1-2-5(3-4)7(6)9(11)13/h1-2,4-7H,3H2/t4-,5+,6-,7-/m1/s1

InChI key

KANQIAARVSWKKG-XZBKPIIZSA-N

General description

Trans-3,6-endomethylene-1,2,3,6-tetrahydrophthaloyl chloride (tNBDC, trans-5-Norbornene-2,3-dicarbonyl chloride) undergoes interfacial polymerization reaction with 1,3-diaminopropane on the surface of a modified polyacrylonitrile (mPAN) during the fabrication of DAPE-SCC/mPAN or DAPE-tNBDC/mPAN TFC membranes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of fine-structure of polyamide thin-film composite membrane under swelling effect by positron annihilation lifetime spectroscopy and molecular dynamics simulation.
Huang YH, et al.
Journal of Membrane Science, 417-418, 201?209-201?209 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service