Skip to Content
Merck
All Photos(1)

Key Documents

111260

Sigma-Aldrich

trans-2-Pentene

99%

Synonym(s):

(E)-2-Pentene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH3
CAS Number:
Molecular Weight:
70.13
Beilstein:
1718795
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010208
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

8.17 psi ( 20 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.379 (lit.)

bp

37 °C (lit.)

mp

−140 °C (lit.)

density

0.649 g/mL at 25 °C (lit.)

SMILES string

CC\C=C\C

InChI

1S/C5H10/c1-3-5-4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

QMMOXUPEWRXHJS-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-Pentene is a significant reactive species in terms of its photochemical consumption and correspondent ozone formation potential[1].

Application

trans-2-Pentene has been used for the calculation of OH levels by the decay method[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Anwar H Khan et al.
Journal of environmental sciences (China), 23(1), 60-64 (2011-04-12)
Night-time OH levels have been determined for UK urban surface environments using two methods, the decay and steady state approximation methods. Measurement data from the UK National Environmental Technology Centre archive for four urban sites (Bristol, Harwell, London Eltham and
Shao Min et al.
Environmental science & technology, 45(2), 514-519 (2010-12-07)
Stringent air-quality control measures were implemented for the 2008 Beijing Olympic Games. This large-scale manmade experiment provided an opportunity to evaluate the effectiveness of measures to reduce the reactivity of hydrocarbons (HCs) from emission sources, which is important for ground-level

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service