Merck
  • Methods for preparing fluorescent and neutral red-stained whole mounts of mouse mammary glands.
Methods for preparing fluorescent and neutral red-stained whole mounts of mouse mammary glands.
Journal of mammary gland biology and neoplasia (2009-11-26)
John D Landua, Adriana P Visbal, Michael T Lewis
ABSTRACT
Whole mount preparations of mouse mammary glands are useful for evaluating overall changes in growth and morphology, and are essential for detecting and evaluating focal or regionally-localized phenotypes that would be difficult to detect or analyze using other techniques. We present three newly developed methods for preparing whole mounts of mammary glands from genetically-engineered mice expressing fluorescent proteins, as well as using either neutral red or a variety of fluorescent dyes. Unlike traditional hematoxylin- or carmine-stained preparations, neutral red-stained and some fluorescent preparations can be used for several common downstream analyses.
Sigma-Aldrich
Neutral Red