Merck
  • Endofin, a novel BMP-SMAD regulator of the iron-regulatory hormone, hepcidin.
Endofin, a novel BMP-SMAD regulator of the iron-regulatory hormone, hepcidin.
Scientific reports (2015-09-12)
Justin B Goh, Daniel F Wallace, Wanjin Hong, V Nathan Subramaniam
ABSTRACT
BMP-SMAD signalling plays a crucial role in numerous biological processes including embryonic development and iron homeostasis. Dysregulation of the iron-regulatory hormone hepcidin is associated with many clinical iron-related disorders. We hypothesised that molecules which mediate BMP-SMAD signalling play important roles in the regulation of iron homeostasis and variants in these proteins may be potential genetic modifiers of iron-related diseases. We examined the role of endofin, a SMAD anchor, and show that knockdown of endofin in liver cells inhibits basal and BMP-induced hepcidin expression along with other BMP-regulated genes, ID1 and SMAD7. We show for the first time, the in situ interaction of endofin with SMAD proteins and significantly reduced SMAD phosphorylation with endofin knockdown, suggesting that endofin modulates hepcidin through BMP-SMAD signalling. Characterisation of naturally occurring SNPs show that mutations in the conserved FYVE domain result in mislocalisation of endofin, potentially affecting downstream signalling and modulating hepcidin expression. In conclusion, we have identified a hitherto unrecognised link, endofin, between the BMP-SMAD signalling pathway, and the regulation of hepcidin expression and iron homeostasis. This study further defines the molecular network involved in iron regulation and provides potential targets for the treatment of iron-related disorders.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Zfyve16
Sigma-Aldrich
MISSION® esiRNA, targeting human ZFYVE16