Merck
  • General approach for electrochemical detection of persistent pharmaceutical micropollutants: Application to acetaminophen.
General approach for electrochemical detection of persistent pharmaceutical micropollutants: Application to acetaminophen.
Biosensors & bioelectronics (2015-05-20)
S Shi, S Reisberg, G Anquetin, V Noël, M C Pham, B Piro
ABSTRACT
We propose in this work a general and versatile methodology for electrochemical monitoring of persistent pharmaceutical micropollutants. The system presented is based on an electroactive and electropolymerized hapten (mimetic molecule of the pollutant to be detected) and a specific antibody that competitively binds either the hapten or the pollutant. The current delivered by the device depends on this competitive equilibrium and therefore on the pollutant's concentration. The determination of the pharmaceutical product operates within minutes, using square wave voltammetry without labeling or addition of a reactant in solution; the competitive hapten/antibody transduction produces a "signal-on" (a current increase). Applied to acetaminophen, this electrochemical immunosensor presents a very low detection limit of ca. 10 pM, (S/N=3) and a very high selectivity towards structural analogs (aspirin, BPA, and piperazine) even in a mixture.
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
2-Pentynoic acid, 97%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture