Merck
  • Isolation and differentiation potential of human mesenchymal stem cells from adipose tissue harvested by water jet-assisted liposuction.
Isolation and differentiation potential of human mesenchymal stem cells from adipose tissue harvested by water jet-assisted liposuction.
Aesthetic surgery journal (2015-05-27)
Juliane Meyer, Achim Salamon, Nicole Herzmann, Stefanie Adam, Hans-Dieter Kleine, Inge Matthiesen, Klaus Ueberreiter, Kirsten Peters
ABSTRACT
In recent years the therapeutic application of extracted adipose tissue for autologous fat grafting and the application of adipose tissue-derived mesenchymal stem cells (adMSC) isolated thereof has progressed. Water-jet assisted liposuction (WAL) is 1 procedure for harvesting adipose tissue and provides a favorable aesthetic outcome combined with high tissue protection. Tissue aspirated by WAL has been successfully applied in grafting procedures. The aims of this study were to confirm the tissue viability and to understand the abundance and mesenchymal differentiation capacity of stem cells within the tissue. We analyzed tissue integrity of WAL tissue particles via fluorescence microscopy. The adMSC content was determined by isolating the cells from the tissue. The mesenchymal differentiation capacity was confirmed with cytochemical staining methods. The stromal vascular fraction of WAL tissue showed high viability and contained an average of 2.6 × 105 CD34-positive cells per milliliter of tissue. Thus WAL tissue contains a high number of stem cells. Furthermore adMSC isolated from WAL tissue showed typical mesenchymal differentiation potential. WAL of adipose tissue is well suited for autologous fat grafting because it retains tissue viability. Furthermore it is a valid source for the subsequent isolation of adMSC with multipotent differentiation potential. 3 Therapeutic.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture