Merck
  • p204-Mediated innate antiviral responses in mouse adipose cells and their effects on cell functions.
p204-Mediated innate antiviral responses in mouse adipose cells and their effects on cell functions.
Immunology and cell biology (2014-10-08)
Lili Yu, Peng Liu, Zhenghui Liu, Weiwei Zhu, Keqin Yan, Qiaoyuan Chen, Daishu Han
ABSTRACT
Viruses can infect adipose tissues. However, innate antiviral responses in adipose cells and their effects on adipocyte function have not yet been intensively investigated. In this study, p204-initiated innate antiviral responses in mouse adipose cells were examined. Cytosolic DNA sensor p204 and its signaling adaptor stimulator of interferon (IFN) genes (STING) were constitutively expressed in primary preadipocytes. Synthetic herpes simplex viral DNA (HSV60), a p204 ligand, induced type I IFN expression by activating IFN regulatory factor 3. Major antiviral proteins, including IFN-stimulating gene 15, 2',5'-oligoadenylate synthetase and Mx GTPase 1, in preadipocytes were upregulated by HSV60. HSV60-triggered innate antiviral responses were significantly reduced by inhibition of p204 signaling with specific small interfering RNA targeting p204 or STING. HSV60 inhibited the differentiation of preadipocytes to mature adipocytes and enhanced the proliferation of adipose cells. Moreover, HSV60 induced innate antiviral responses in mature adipocytes and inhibited expressions of several adipokines, including leptin, adiponectin and resistin. These results indicated that p204 initiated innate antiviral responses in adipose cells, thereby modulating adipocyte function.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Anti-IFI204 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ZBP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)