Merck
  • Recurrent SKIL-activating rearrangements in ETS-negative prostate cancer.
Recurrent SKIL-activating rearrangements in ETS-negative prostate cancer.
Oncotarget (2015-03-10)
Matti Annala, Kati Kivinummi, Joonas Tuominen, Serdar Karakurt, Kirsi Granberg, Leena Latonen, Antti Ylipää, Liisa Sjöblom, Pekka Ruusuvuori, Outi Saramäki, Kirsi M Kaukoniemi, Olli Yli-Harja, Robert L Vessella, Teuvo L J Tammela, Wei Zhang, Tapio Visakorpi, Matti Nykter
ABSTRACT
Prostate cancer is the third most common cause of male cancer death in developed countries, and one of the most comprehensively characterized human cancers. Roughly 60% of prostate cancers harbor gene fusions that juxtapose ETS-family transcription factors with androgen regulated promoters. A second subtype, characterized by SPINK1 overexpression, accounts for 15% of prostate cancers. Here we report the discovery of a new prostate cancer subtype characterized by rearrangements juxtaposing the SMAD inhibitor SKIL with androgen regulated promoters, leading to increased SKIL expression. SKIL fusions were found in 6 of 540 (1.1%) prostate cancers and 1 of 27 (3.7%) cell lines and xenografts. 6 of 7 SKIL-positive cancers were negative for ETS overexpression, suggesting mutual exclusivity with ETS fusions. SKIL knockdown led to growth arrest in PC-3 and LNCaP cell line models of prostate cancer, and its overexpression led to increased invasiveness in RWPE-1 cells. The role of SKIL as a prostate cancer oncogene lends support to recent studies on the role of TGF-β signaling as a rate-limiting step in prostate cancer progression. Our findings highlight SKIL as an oncogene and potential therapeutic target in 1-2% of prostate cancers, amounting to an estimated 10,000 cancer diagnoses per year worldwide.
Product Description
Supelco
N,N′-Bis(acryloyl)cystamine, BioReagent, suitable for electrophoresis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%