Merck
  • A double-blinded, prospective study to define antigenemia and quantitative real-time polymerase chain reaction cutoffs to start preemptive therapy in low-risk, seropositive, renal transplanted recipients.
A double-blinded, prospective study to define antigenemia and quantitative real-time polymerase chain reaction cutoffs to start preemptive therapy in low-risk, seropositive, renal transplanted recipients.
Transplantation (2014-05-21)
Elias David-Neto, Ana H K Triboni, Flavio J Paula, Lucy S Vilas Boas, Clarisse M Machado, Fabiana Agena, Acram Z A Latif, Cecília S Alencar, Ligia C Pierrotti, William C Nahas, Helio H Caiaffa-Filho, Claudio S Pannuti
ABSTRACT
Cytomegalovirus (CMV) disease occurs in 16% to 20% of low-risk, CMV-positive renal transplant recipients. The cutoffs for quantitative real-time polymerase chain reaction (qPCR) or phosphoprotein (pp65) antigenemia (pp65emia) for starting preemptive therapy have not been well established. We measured qPCR and pp65emia weekly from day 7 to day 120 after transplantation, in anti-CMV immunoglobulin G–positive donor and recipient pairs. Patients and physicians were blinded to the test results. Suspicion of CMV disease led to the order of new tests. In asymptomatic viremic patients, the highest pp65emia and qPCR values were used, whereas we considered the last value before diagnosis in those with CMV disease. We collected a total of 1,481 blood samples from 102 adult patients. Seventeen patients developed CMV disease, 54 presented at least one episode of viremia that cleared spontaneously, and 31 never presented viremia. Five patients developed CMV disease after the end of the study period. The median (95% confidence interval) pp65emia and qPCR values were higher before CMV disease than during asymptomatic viremia (6 [9–82] vs. 3 [1–14] cells/10(6) cells; P<0.001 and 3,080 [1,263–15,605] vs. 258 [258–1,679] copies/mL; P=0.008, respectively). The receiver operating characteristic curve showed that pp65emia 4 cells/10(6) cells or greater showed a sensitivity and specificity to predict CMV disease of 69% and 81%, respectively (area, 0.769; P=0.001), with a positive predictive value of 37% and a negative predictive value of 93%. For qPCR 2,000 copies/mL or higher, the positive predictive value and negative predictive value were 57% and 91%, respectively (receiver operating characteristic area, 0.782; P=0.000). With these cutoffs, both methods are appropriate for detecting CMV disease.
Sigma-Aldrich
Acetone, purum, ≥99.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%