Merck
  • Behaviour of endothelial cells in a tridimensional in vitro environment.
Behaviour of endothelial cells in a tridimensional in vitro environment.
BioMed research international (2015-03-20)
Raif Eren Ayata, Stéphane Chabaud, Michèle Auger, Roxane Pouliot
ABSTRACT
Angiogenesis is a fundamental process in healing, tumor growth, and a variety of medical conditions. For this reason, in vitro angiogenesis is an area of interest for researchers. Additionally, in vitro angiogenesis is important for the survival of prevascularized tissue-engineering models. The aim of this study was to observe the self-tubular organization behaviour of endothelial cells in the self-assembly method. In this study, bilayered and dermal substitutes were prepared using the self-assembly method. Histological, immunostaining, and biochemical tests were performed. The behavioural dynamics of endothelial cells in this biological environment of supportive cells were observed, as were the steps of the in vitro angiogenic cascade with self-organizing capillary-like structures formation. The epidermal component of the substitutes was seen to promote network expansion and density. It also increased the quantity of angiogenic factors (VEGF and Ang-1) without increasing the proinflammatory factor (IL-8). In addition, the increased MMP activity contributed to matrix degradation, which facilitated capillary formation.
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acetone, purum, ≥99.0% (GC)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%