Merck
  • Methylation of PLCD1 and adenovirus-mediated PLCD1 overexpression elicits a gene therapy effect on human breast cancer.
Methylation of PLCD1 and adenovirus-mediated PLCD1 overexpression elicits a gene therapy effect on human breast cancer.
Experimental cell research (2015-02-07)
Haixi Mu, Na Wang, Lijuan Zhao, Shuman Li, Qianqian Li, Ling Chen, Xinrong Luo, Zhu Qiu, Lili Li, Guosheng Ren, Yongzhu Xu, Xiangyang Zhou, Tingxiu Xiang
ABSTRACT
Our previous study showed that PLCD1 significantly decreases cell proliferation and affects cell cycle progression in breast cancer cells. In the present study, we aimed to investigate its functional and molecular mechanisms, and whether or not can become a new target for gene therapies. We found reduced PLCD1 protein expression in breast tumor tissues compared with paired surgical margin tissues. PLCD1 promoter CpG methylation was detected in 55 of 96 (57%) primary breast tumors, but not in surgical-margin tissues and normal breast tissues. Ectopic expression of PLCD1 inhibited breast tumor cell proliferation in vivo by inducing apoptosis and suppressed tumor cell migration by regulating cytoskeletal reorganization proteins including RhoA and phospho-cofilin. Furthermore, we found that PLCD1 induced p53 accumulation, increased p27 and p21 protein levels, and cleaved PARP. Finally, we constructed an adenoviral vector expressing PLCD1 (AdH5-PLCD1), which exhibited strong cytotoxicity in breast cancer cells. Our findings provide insights into the development of PLCD1 gene therapies for breast cancer and perhaps, other human cancers.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-PLCD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Pricing and availability is not currently available.