Merck
  • Sulfhydration of p66Shc at cysteine59 mediates the antioxidant effect of hydrogen sulfide.
Sulfhydration of p66Shc at cysteine59 mediates the antioxidant effect of hydrogen sulfide.
Antioxidants & redox signaling (2014-04-29)
Zhi-Zhong Xie, Mei-Mei Shi, Li Xie, Zhi-Yuan Wu, Guang Li, Fei Hua, Jin-Song Bian
ABSTRACT
Mitochondrion is considered as the major source of intracellular reactive oxygen species (ROS). H2S has been reported to be an antioxidant, but its mechanism remains largely elusive. P66Shc is an upstream activator of mitochondrial redox signaling. The aim of this study was to explore whether the antioxidant effect of H2S is mediated by p66Shc. Application of exogenous H2S with its donor, NaHS, or overexpression of its generating enzyme, cystathionine β-synthase, induced sulfhydration of p66Shc, but inhibited its phosphorylation caused by H2O2/D-galactose in SH-SY5Y cells or in the mice cortex. H2S also decreased mitochondrial ROS production and protected neuronal cells against stress-induced senescence. PKCβII and PP2A are the two key proteins to regulate p66Shc phosphorylation. Although H2S failed to affect the activities of these two proteins, it disrupted their association. Cysteine-59 resides in proximity to serine-36, the phosphorylation site of p66Shc. The C59S mutant attenuated the above-described biological function of H2S. We revealed a novel mechanism for the antioxidant effect of H2S and its role in oxidative stress-related diseases. H2S inhibits mitochondrial ROS production via the sulfhydration of Cys-59 residue, which in turn, prevents the phosphorylation of p66Shc.
Product Description
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%