Merck
  • Apoptotic effects of 7,8-dihydroxyflavone in human oral squamous cancer cells through suppression of Sp1.
Apoptotic effects of 7,8-dihydroxyflavone in human oral squamous cancer cells through suppression of Sp1.
Oncology reports (2014-12-02)
Ra Ham Lee, Jae-Cheon Shin, Ka-Hwi Kim, Yung Hyun Choi, Jung-Il Chae, Jung-Hyun Shim
ABSTRACT
7,8-Dihydroxyflavone (7,8-DHF) is a member of the flavonoid family and has recently been identified as a brain-derived neurotrophic factor mimetic that selectively activates tropomyosin-receptor kinase B with high affinity. The antioxidant and anticancer effects of 7,8-DHF have been reported. However, the pharmacological mechanisms of 7,8-DHF in oral cancer are unclear. Thus, we investigated the mechanisms of the antiproliferative action of 7,8-DHF on HN22 and HSC4 oral squamous cell carcinoma cell lines. We demonstrated that 7,8-DHF decreased cell growth and induced apoptosis in the HN22 and HSC4 cells through regulation of specificity protein 1 (Sp1) using the MTS assay, DAPI staining, Annexin V, propidium iodide staining, reverse transcription-polymerase chain reaction, immunocytochemistry, pull-down assay and western blot analysis. The results showed that the Sp1 protein bound with 7,8-DHF in the HN22 and HSC4 cells. Taken together, the results suggest that 7,8-DHF could modulate Sp1 transactivation and induce apoptotic cell death by regulating the cell cycle and suppressing antiapoptotic proteins. Furthermore, 7,8-DHF may be valuable for cancer prevention and better clinical outcomes.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
DL-Dithiothreitol solution, 1 M in H2O