Merck
  • Prophylactic and therapeutic effects of Acanthopanax senticosus Harms extract on murine collagen-induced arthritis.
Prophylactic and therapeutic effects of Acanthopanax senticosus Harms extract on murine collagen-induced arthritis.
Phytotherapy research : PTR (2014-05-07)
Yusuke Takahashi, Maki Tanaka, Ryosei Murai, Kageaki Kuribayashi, Daisuke Kobayashi, Nozomi Yanagihara, Naoki Watanabe
ABSTRACT
Evidences are accumulating that extract of Acanthopanax senticosus Harms (ASH; syn Eleutherococcus senticosus [Rupr. & Maxim.] Maxim), a shrub native to Northeastern Asia, has antiinflammatory effects. In this study, we examined prophylactic and therapeutic effects of ASH extract (ASHE) on rheumatoid arthritis using collagen-induced arthritis (CIA) mouse model. Acanthopanax senticosus Harms extract was administered before the onset of arthritis in the prophylaxis model. In the therapeutic model, ASHE was administered after the onset of arthritis with or without anti-TNF-α antibody. The ASHE treatment showed efficacy before onset of CIA but there was no effect after CIA was established. The ASHE treatment delayed the onset and decreased severity of CIA. In vitro examinations showed that ASHE is an antioxidant and that ASHE suppresses TNF-α and interleukin-6 production in human peripheral blood mononuclear cells. The combination therapy with ASHE and anti-TNF-α antibody reduced the severity of arthritis compared with anti-TNF-α antibody alone. The present study shows that ASHE has prophylactic effect against CIA and support therapeutic effect of anti-TNF-α antibody.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Riboflavin (B2), analytical standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard