Merck
  • Synthesis and antioxidant activity of nitrohydroxytyrosol and its acyl derivatives.
Synthesis and antioxidant activity of nitrohydroxytyrosol and its acyl derivatives.
Journal of agricultural and food chemistry (2014-09-30)
Mariana Trujillo, Elena Gallardo, Andrés Madrona, Laura Bravo, Beatriz Sarriá, José A González-Correa, Raquel Mateos, José Luis Espartero
ABSTRACT
A series of nitroderivatives has been synthesized from hydroxytyrosol, the natural olive oil phenol, to increase the assortment of compounds with putative effects against Parkinson's disease. Nitrohydroxytyrosyl esters were obtained from nitrohydroxytyrosol using a chemoselective one-step, high-yield, transesterification procedure. The antioxidant activity of these new series of nitrocatechols was evaluated using FRAP, ABTS, and ORAC assays and compared to that of free hydroxytyrosol. The nitro functional group induced a significant increase in the antioxidant activity of nitrohydroxytyrosol compared to hydroxytyrosol. Regarding nitroester derivatives, variable antioxidant activity was observed depending on the acyl side-chain length; shorter chains maintained or even enhanced the antioxidant activity compared to nitrohydroxytyrosol, decreasing the activity with longer side chains in keeping with their lipophilic nature. Therefore, it may be concluded that nitroester derivatives of hydroxytyrosol, which may be obtained by a simple, high-yield reaction, have elevated antioxidant activity and thus present potential bioactivity.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, purum, ≥99.0% (GC)