Merck
  • Bevacizumab reduces neurocan content and gene expression in newborn rat retina in vitro.
Bevacizumab reduces neurocan content and gene expression in newborn rat retina in vitro.
Investigative ophthalmology & visual science (2014-07-26)
Paloma G Krempel, Monique Matsuda, Mônica V Marquezini, Thayane G Seixas, Grasiella M Ventura, Alfred Sholl-Franco, Nádia C O Miguel, Mário L R Monteiro
ABSTRACT
Extracellular matrix (ECM) and cellular membrane proteoglycans (PGs) play important roles in neural differentiation and cell adhesion. Vascular endothelial growth factor, an important signal protein in vascular and retinal neural cell development, is retained in the ECM due to its high affinity for PG. Bevacizumab, an anti-VEGF agent, has been extensively used for treating retinal diseases in adult and newborn patients, although its effect on the developing retina remains largely unknown. The purpose of this study was to investigate the effect of bevacizumab on neurocan, phosphacan, and syndecan-3 PG levels in newborn rat retina. Retinal explants of sixty 2-day-old Lister hooded rats were obtained after eye enucleation and maintained in culture media with or without bevacizumab for 48 hours. Immunohistochemical staining was assessed against neurocan, phosphacan, and syndecan-3. Proteoglycan content was quantified based on the intensity of immunohistochemical labeling. Gene expressions were quantified by real-time reverse-transcription polymerase chain reaction. The results from the treatment and control groups were compared. No significant difference in the staining intensity and mRNA expression of phosphacan and syndecan-3 was observed between the groups. However, a significant decrease in neurocan content and mRNA expression was observed in bevacizumab-treated retinal explants compared with controls. Bevacizumab did not affect phosphacan and syndecan-3 levels but decreased neurocan content and gene expression. Therefore, it may interfere with early postnatal retinal cell differentiation. Although further studies are necessary to confirm our findings, we suggest anti-VEGF agents be used with caution in developing retinal tissue.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
SAFC
HEPES
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water