Merck
  • The 15-lipoxygenase inhibitory, antioxidant, antimycobacterial activity and cytotoxicity of fourteen ethnomedicinally used African spices and culinary herbs.
The 15-lipoxygenase inhibitory, antioxidant, antimycobacterial activity and cytotoxicity of fourteen ethnomedicinally used African spices and culinary herbs.
Journal of ethnopharmacology (2014-08-26)
Jean Paul Dzoyem, Victor Kuete, Lyndy J McGaw, Jacobus N Eloff
ABSTRACT
Culinary herbs and spices are widely used ethnomedically across Africa. They are traditionally employed in the treatment of several ailments including inflammation disorders, pain alleviation and infectious diseases. Pharmacological studies are necessary to provide a scientific basis to substantiate their traditional use and safety. In this study, the 15-lipoxygenase inhibitory, antioxidant, antimycobacterial and the cytotoxic activities, total phenolic and flavonoid contents of fourteen edible plants were investigated. The 15-lipoxygenase inhibitory activity was evaluated by the ferrous oxidation-xylenol orange (FOX) assay method. The antioxidant activity was determined using free-radical scavenging assays. The antimycobacterial activity was determined by a broth microdilution method against three species of mycobacteria: Mycobacterium smegmatis, Mycobacterium aurum and Mycobacterium fortuitum using tetrazolium violet as growth indicator. The cytotoxicity was assessed by the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay on Vero monkey kidney cells. All the extracts tested had some 15-lipoxygenase inhibitory activity ranging from 32.9 to 78.64%. Adansonia digitata (fruit) had the highest antioxidant capacity (IC₅₀ values of 8.15 μg/mL and 9.16 μg/mL in the DPPH and ABTS assays respectively; TEAC of 0.75 in the FRAP assay) along with the highest amount of total phenolics (237.68 mg GAE/g) and total flavonoids (16.14 mg E/g). There were good correlations between DPPH and ABTS values (R(2) 0.98) and between total phenolics and total flavonoids (R(2) 0.94). Tamarindus indica had significant antimycobacterial activity against Mycobacterium aurum (MIC 78 μg/mL). As could be expected with edible plants, all the extracts had a relatively low cytotoxicity with LC₅₀ values higher than 102 μg/mL with the exception of the two Aframomum species (33 and 74 μg/mL). This study provides scientific support for some of the the traditional uses and the pharmacological activities of the culinary herbs and spices investigated. The results suggest that increasing intake of some of these herbs may be useful in preventing or reducing the progression of lifestyle-related diseases. The diversity of the pharmacological activities of the extract from the fruit of Adansonia digitata suggested that this plant might be valuable for application in human and animal health.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Acetone, purum, ≥99.0% (GC)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material