Merck
  • Thyrostimulin deficiency does not alter peripheral responses to acute inflammation-induced nonthyroidal illness.
Thyrostimulin deficiency does not alter peripheral responses to acute inflammation-induced nonthyroidal illness.
American journal of physiology. Endocrinology and metabolism (2014-08-15)
Clementine J J van Zeijl, Olga V Surovtseva, Joan Kwakkel, Hermina C van Beeren, J H Duncan Bassett, J H Duncan Bassett, Graham R Williams, Wilmar M Wiersinga, Eric Fliers, Anita Boelen
ABSTRACT
Thyrostimulin, a putative glycoprotein hormone, comprises the subunits GPA2 and GPB5 and activates the TSH receptor (TSHR). The observation that proinflammatory cytokines stimulate GPB5 transcription suggested a role for thyrostimulin in the pathogenesis of nonthyroidal illness syndrome (NTIS). In the present study, we induced acute inflammation by LPS administration to GPB5(-/-) and WT mice to evaluate the role of thyrostimulin in peripheral thyroid hormone metabolism during NTIS. In addition to serum thyroid hormone concentrations, we studied mRNA expression and activity of deiodinase types I, II, and III (D1, D2, and D3) in peripheral T3 target tissues, including liver, muscle, and white and brown adipose tissue (WAT and BAT), of which the latter three express the TSHR. LPS decreased serum free (f)T4 and fT3 indexes to a similar extent in GPB5(-/-) and WT mice. Serum reverse (r)T3 did not change following LPS administration. LPS also induced significant alterations in tissue D1, D2, and D3 mRNA and activity levels, but only the LPS-induced increase in WAT D2 mRNA expression differed between GPB5(-/-) and WT mice. In conclusion, lacking GPB5 during acute illness does not affect the LPS-induced decrease of serum thyroid hormones while resulting in subtle changes in tissue D2 expression that are unlikely to be mediated via the TSHR.
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
cAMP Enzyme Immunoassay Kit, Direct
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine