Merck
  • Safety and technological characterization of Staphylococcus equorum isolates from jeotgal, a Korean high-salt-fermented seafood, for starter development.
Safety and technological characterization of Staphylococcus equorum isolates from jeotgal, a Korean high-salt-fermented seafood, for starter development.
International journal of food microbiology (2014-08-12)
Do-Won Jeong, Seulhwa Han, Jong-Hoon Lee
ABSTRACT
To select starters for jeotgal, a traditional Korean high-salt-fermented seafood, the safety and technological properties of its predominant bacteria isolates, which were identified as Staphylococcus equorum, were assessed. Among the 185 S. equorum isolates from jeotgal, 126 ampicillin-sensitive strains were subjected to assessments for antibiotic susceptibility and safety hazards. Sixty-six out of the 126 S. equorum strains exhibited phenotypic resistances to at least one antibiotic, and their prevailing resistances were to penicillin G (34.1%), erythromycin (9.5%) and trimethoprim (9.5%). Twenty-four S. equorum strains expressed resistance to at least two antibiotics. The lnuA for lincomycin (four strains) and pbp for β-lactam (three strains) were amplified by PCR. α-Hemolytic activity was not detected from the 126 strains, and 87 strains presented δ-hemolytic activity. Among the 87 strains, three strains exhibited β-hemolytic activity. Thirty-seven strains formed a biofilm. A hemolysin gene homologous to that of Staphylococcus epidermidis was amplified from an S. equorum strain with β-hemolytic activity by PCR; however, no PCR product homologous to the previously known staphylococcal enterotoxin genes was amplified. Thirty-nine S. equorum strains cleared all of the tested safety hazards and were adopted for technological property assessments. Among these strains, 16 strains exhibited protease, lipase and nitrate reductase activities, and seven strains did not produce four types of biogenic amines. Five biogenic amine non-producers exhibited three enzyme activities. Most of the strains could grow on the agar with 20% NaCl, and 13 strains maintained growth at the 25% NaCl condition. S. equorum KS1039, which is the most applicable strain that covers the safety and technological requirements for jeotgal, can grow at the 25% NaCl condition. Through this research study, we reconfirmed the necessity of characterization in the functionality and safety of S. equorum for starter development because all of the tested phenotypic characteristics were expressed in strain-specific manners.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Trimethoprim, European Pharmacopoeia (EP) Reference Standard