Merck
The role of arginine methylation in the DNA damage response.
DNA repair (2013-05-21)
Yannick Auclair, Stéphane Richard
ABSTRACT
Post-translational modifications are well-known modulators of DNA damage signaling and epigenetic gene expression. Protein arginine methylation is a covalent modification that results in the addition of methyl groups to the nitrogen atoms of the arginine side chains and is catalyzed by a family of protein arginine methyltransferases (PRMTs). In the past, arginine methylation was mainly observed on abundant proteins such as RNA-binding proteins and histones, but recent advances have revealed a plethora of arginine methylated proteins implicated in a variety of cellular processes including RNA metabolism, epigenetic regulation and DNA repair pathways. Herein, we discuss these recent advances, focusing on the role of PRMTs in DNA damage signaling and its importance for maintaining genomic stability.
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
SAFC
L-Arginine
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Arginine, European Pharmacopoeia (EP) Reference Standard